Vinyltrimethoxysilane 97% stabilized

Supplier: Ambeed
Danger

Synonyms: Trimethoxy(vinyl)silane

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A754635-25G
76776-228EA 6.37 USD
76776-228
Vinyltrimethoxysilane 97% stabilized
Vinyltrimethoxysilane

Caution: for Research Use Only

Formula: C₅H₁₂O₃Si
MW: 148.23 g/mol
MDL Number: MFCD00008605
CAS Number: 2768-02-7
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR