4-Fluorophenyl isocyanate 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene
A655096-5G
A655096-1G
A655096-25G
76775-582EA
17
USD
76775-582
77573-102
77878-480
4-Fluorophenyl isocyanate 97%
4-Fluorophenyl isocyanate
Caution: for Research Use Only
Formula:
C₇H₄FNO MW: 137.11 g/mol |
MDL Number:
MFCD00002023 CAS Number: 1195-45-5 UN: 2478 ADR: 3,6.1 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...