Chromone-2-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Oxo-4H-1-benzopyran-2-carboxylic acid, Chromocarb, 4-Oxo-4H-chromene-2-carboxylic acid
A526233-10G
A526233-1G
A526233-250MG
A526233-25G
A526233-25MG
A526233-5G
A526233-100G
76774-684EA
28
USD
76774-684
76774-686
76774-688
76774-690
76774-692
76774-694
77627-074
Chromone-2-carboxylic acid 98%
Chromone-2-carboxylic Acid
Caution: for Research Use Only
Formula:
C₁₀H₆O₄ |
MDL Number:
MFCD00006838 CAS Number: 4940-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...