5-Carboxytetramethylrhodamine (5-TAMRA) 98%

Supplier: Ambeed
Warning

Synonyms: 5-TAMRA, 5-Carboxytetramethylrhodamine, 2-(3,6-Bis(dimethylamino)xanthylium-9-yl)-5-carboxybenzoate, 2-[3,6-Bis(dimethylamino)xanthen-10-ium-9-yl]-5-carboxybenzoate, TAMRA, 5-isomer

A505764-100MG A505764-10MG A505764-1G A505764-1MG A505764-250MG A505764-25MG A505764-5G A505764-5MG A505764-25G
76774-530EA 65 USD
76774-530 76774-532 76774-534 76774-536 76774-538 76774-540 76774-542 76774-544 77878-289
5-Carboxytetramethylrhodamine (5-TAMRA) 98%
5-Carboxytetramethylrhodamine (5-TAMRA)

Caution: for Research Use Only

Formula: C₂₅H₂₂N₂O₅
MDL Number: MFCD00269769
CAS Number: 91809-66-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR