N,N-Diphenylcarbamoyl chloride 98% (by GC)

Supplier: Ambeed
Danger

A361611-25G A361611-5G A361611-100G
76773-348EA 14 USD
76773-348 76773-350 77626-698
N,N-Diphenylcarbamoyl chloride 98% (by GC)
N,N-Diphenylcarbamoyl chloride

Caution: for Research Use Only

Formula: C₁₃H₁₀ClNO
MDL Number: MFCD00000633
CAS Number: 83-01-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR