6-Fluoro-m-toluidine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-4-fluorotoluene, 6-Fluoro-m-methylaniline, 2-Fluoro-5-methylaniline

A305193-100G A305193-10G A305193-25G A305193-500G A305193-5G
76772-700EA 40 USD
76772-700 76772-702 76772-704 76772-706 76772-708
6-Fluoro-m-toluidine 98%
6-Fluoro-m-toluidine

Caution: for Research Use Only

Formula: C₇H₈FN
MW: 125.15 g/mol
MDL Number: MFCD00007654
CAS Number: 452-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR