(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%

Supplier: Ambeed
Warning

Synonyms: (S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinemethanol, (S)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol, (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline

A300945-100G A300945-10G A300945-1G A300945-250MG A300945-25G A300945-5G
76772-630EA 520 USD
76772-630 76772-632 76772-634 76772-636 76772-638 76772-640
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol

Caution: for Research Use Only

Formula: C₁₀H₁₃NO
MDL Number: MFCD01631316
CAS Number: 18881-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR