(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-(-)-1,2,3,4-Tetrahydro-3-isoquinolinemethanol, (S)-(1,2,3,4-Tetrahydroisoquinolin-3-yl)methanol, (S)-3-Hydroxymethyl-1,2,3,4-tetrahydroisoquinoline
A300945-100G
A300945-10G
A300945-1G
A300945-250MG
A300945-25G
A300945-5G
76772-630EA
520
USD
76772-630
76772-632
76772-634
76772-636
76772-638
76772-640
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol 97%
(S)-1,2,3,4-Tetrahydroisoquinoline-3-methanol
Caution: for Research Use Only
Formula:
C₁₀H₁₃NO |
MDL Number:
MFCD01631316 CAS Number: 18881-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...