Boc-trans-4-hydroxy-L-proline 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline
A273613-1000G
A273613-100G
A273613-10G
A273613-1G
A273613-500G
A273613-5G
A273613-25G
76772-184EA
245
USD
76772-184
76772-186
76772-188
76772-190
76772-192
76772-194
77092-872
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline
Caution: for Research Use Only
Formula:
C₁₀H₁₇NO₅ MW: 231.3 g/mol |
MDL Number:
MFCD00053370 CAS Number: 13726-69-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...