Boc-trans-4-hydroxy-L-proline 97%

Supplier: Ambeed
Warning

Synonyms: (2S,4R)-1-(tert-Butoxycarbonyl)-4-hydroxypyrrolidine-2-carboxylic acid, Boc-Hyp-OH, trans-N-Boc-4-hydroxy-L-proline

A273613-1000G A273613-100G A273613-10G A273613-1G A273613-500G A273613-5G A273613-25G
76772-184EA 245 USD
76772-184 76772-186 76772-188 76772-190 76772-192 76772-194 77092-872
Boc-trans-4-hydroxy-L-proline 97%
Boc-trans-4-hydroxy-L-proline

Caution: for Research Use Only

Formula: C₁₀H₁₇NO₅
MW: 231.3 g/mol
MDL Number: MFCD00053370
CAS Number: 13726-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR