2-Fluoro-4-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A234913-100G A234913-10G A234913-1G A234913-250MG A234913-25G A234913-5G
76771-236EA 182 USD
76771-236 76771-238 76771-240 76771-242 76771-244 76771-246
2-Fluoro-4-(trifluoromethyl)benzonitrile 98%
2-Fluoro-4-(trifluoromethyl)benzonitrile

Caution: for Research Use Only

Formula: C₈H₃F₄N
MDL Number: MFCD00061279
CAS Number: 146070-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR