7-Bromoindole 97%
Supplier: Ambeed
|
Warning
|
A211565-100G
A211565-1G
A211565-5G
103818-114EA
17
USD
103818-114
103848-808
76770-526
77087-220
77087-222
7-Bromoindole 97%
7-Bromoindole
Caution: for Research Use Only
Formula:
C₈H₆BrN Boiling Pt: 316.9 °C (760 mm Hg) Melting Pt: 41…44 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00799492 CAS Number: 51417-51-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...