5-Amino-2-chloro-4-nitrobenzotrifluoride 95%

Supplier: Ambeed
Warning

Synonyms: 2-Nitro-4-chloro-5-(trifluoromethyl)aniline, 5-Amino-2-chloro-4-nitrobenzotrifluoride

A200707-1G
76770-138EA 19.1 USD
76770-138
5-Amino-2-chloro-4-nitrobenzotrifluoride 95%
5-Amino-2-chloro-4-nitrobenzotrifluoride

Caution: for Research Use Only

Formula: C₇H₄ClF₃N₂O₂
MW: 240.5700 g/mol
MDL Number: MFCD03094264
CAS Number: 167415-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR