2-Bromo-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A186424-100G A186424-10G A186424-1G A186424-25G A186424-500G A186424-5G
76769-756EA 138 USD
76769-756 76769-758 76769-760 76769-762 76769-764 76769-766
2-Bromo-3-nitrobenzoic acid 98%
2-Bromo-3-nitrobenzoic acid

Caution: for Research Use Only

Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
MDL Number: MFCD00074899
CAS Number: 573-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR