5-Bromo-2-iodotoluene 98%
Supplier: Ambeed
|
Warning
|
A186165-100G
A186165-10G
A186165-500G
A186165-25G
76769-740EA
55
USD
76769-740
76769-742
76769-744
77084-150
5-Bromo-2-iodotoluene 98%
5-Bromo-2-iodotoluene
Caution: for Research Use Only
Formula:
C₇H₆BrI |
MDL Number:
MFCD00060664 CAS Number: 116632-39-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...