Benzethonium chloride 98%

Supplier: Ambeed
Warning

Synonyms: Hyamine® 1622, N-Benzyl-N,N-dimethyl-2-(2-(4-(2,4,4-trimethylpentan-2-yl)phenoxy)ethoxy)ethanaminium chloride

A182907-100MG
76769-614EA 5.31 USD
76769-614
Benzethonium chloride 98%
Benzethonium chloride

Caution: for Research Use Only

Formula: C₂₇H₄₂ClNO₂
MW: 448.09 g/mol
MDL Number: MFCD00011742
CAS Number: 121-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR