Methyl-6-formylpicolinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 6-formylpyridine-2-carboxylate, Methyl 6-formyl-2-pyridinecarboxylate

A164550-10G A164550-250MG A164550-5G
103805-878EA 13 USD
103805-878 103844-134 76768-756 76768-758 77320-312
Methyl-6-formylpicolinate 98%
Methyl 6-formylpicolinate

Caution: for Research Use Only

Formula: C₈H₇NO₃
MW: 165.1500 g/mol
Boiling Pt: 296.6 °C (760 mm Hg)
Melting Pt: 101…104 °C
Storage Temperature: Freezer
MDL Number: MFCD07367925
CAS Number: 69950-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR