N-Boc-ethylenediamine 97%

Supplier: Ambeed
Danger

A144595-10G A144595-5G A144595-25G
103841-500EA 5 USD
103841-500 76767-958 77299-400 77316-752
N-Boc-ethylenediamine 97%
N-Boc-ethylenediamine

Caution: for Research Use Only

Formula: C₇H₁₆N₂O₂
Boiling Pt: 252.7 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191871
CAS Number: 57260-73-8
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR