2-Fluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
o-Fluorobenzoic acid
A129612-1000G
A129612-100G
A129612-10G
A129612-25G
A129612-500G
76767-132EA
63
USD
76767-132
76767-134
76767-136
76767-138
76767-140
2-Fluorobenzoic acid 98%
2-Fluorobenzoic acid
Caution: for Research Use Only
Formula:
C₇H₅FO₂ |
MDL Number:
MFCD00002405 CAS Number: 445-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...