3-Fluoro-4-hydroxybenzaldehyde 97%

Supplier: Ambeed
Warning

A101466-25G A101466-5G A101466-1G A101466-10G A101466-100G
103821-672EA 22 USD
103821-672 76765-176 77282-750 77570-108 77623-824 77876-783
3-Fluoro-4-hydroxybenzaldehyde 97%
3-Fluoro-4-hydroxybenzaldehyde

Caution: for Research Use Only

Formula: C₇H₅FO₂
Boiling Pt: 225.6 °C (760 mm Hg)
Melting Pt: 121…124 °C
Flash Pt: 90.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016626
CAS Number: 405-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR