Methyl-4-bromo-3-nitrobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromobenzoic acid methyl ester, Methyl 4-bromo-3-nitrobenzoate
A101448-100G
A101448-10G
A101448-25G
A101448-1G
A101448-5G
103795-978EA
6
USD
103795-978
103821-662
76765-170
76765-172
76765-174
77282-738
77297-104
Methyl-4-bromo-3-nitrobenzoate 97%
Methyl 4-bromo-3-nitrobenzoate
Caution: for Research Use Only
Formula:
C₈H₆BrNO₄ Boiling Pt: 320.9 °C (760 mm Hg) Melting Pt: 102…103 °C Flash Pt: 147.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00553089 CAS Number: 2363-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...