Thermo Scientific™ TSV Manual Defrost Laboratory Freezer
Supplier: Thermo Fisher Scientific
TSV18FPSA
76764-992EA 3810.85 USD
76764-992
Thermo Scientific™ TSV Manual Defrost Laboratory Freezer
Freezers Laboratory / General Purpose Freezers
The Thermo Scientific™ TSV 18 cu. ft. manual defrost laboratory freezer offers an economical and environmentally friendly solution for safe and reliable general-purpose cold storage. This easy to use freezer with its intuitive design is ideal for a wide range of customers - from small laboratories to large-scale research facilities.
  • Laboratory-grade –20 °C manual defrost freezer
  • Temperature rating –22 to –10 °C (–7.6 to 14 °F)
  • Digital temperature display and adjustable temperature control
  • Audible and visual alarm systems
  • Significant New Alternatives Policy (SNAP)
  • Compliant hydrocarbon refrigerants (R290) and foam
  • Keyed door lock
  • 2 adjustable feet in front and 2 rollers at the rear
  • Easy-to-clean, durable finish
The full-size cabinet provides ample freezing space, while the digital controller and keyed locking features offer confidence and security. The Thermo Scientific TSV manual defrost freezer is thoughtfully designed for a variety of laboratory needs, environmentally and energy efficient, all while remaining economically priced.
Certifications: cETLus certified and Nationally Recognized Testing; Laboratory (NRTL) listed.