VACUUBRAND® Chemistry-HYBRID Vacuum Pump
Supplier: Brandtech
20798563
76764-990EA 9548.15 USD
76764-990
VACUUBRAND® Chemistry-HYBRID Vacuum Pump
Pumps Vacuum Diaphragm Pumps
Hybrid vacuum pump provides extensive pumping capabilities for solvent vapors along with enhanced opposition to aggressive gases.
  • Minimized internal corrosion
  • Reduction in oil changes
  • Telescopic design
The RC 6 chemistry-Hybrid pump combines the technology of a two-stage rotary vane pump with a two-stage chemistry diaphragm pump. Partial pressures of solvent vapors are kept at levels below their condensation points, reducing oxygen and corrosive gas concentrations.
In most practical operations, a cold trap is often unnecessary making this both economical and easy to maintain.
Ordering information: Two year warranty.
Delivery information: Supplied with operating manual and US plug.
Packaging: Recyclable packaging.