Anti-RNA-directed RNA polymerase Rabbit Polyclonal Antibody

Supplier: Prosci
11-035
76757-202EA 495 USD
76757-202
Anti-RNA-directed RNA polymerase Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR