Anti-EMD Rabbit Polyclonal Antibody

Supplier: Prosci
18-218
76751-006EA 372.5 USD
76751-006
Anti-EMD Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EMD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR