n-Hexacosane 98%

Supplier: Ambeed
Warning

A953813-5G
76729-330EA 39 USD
76729-330
n-Hexacosane 98%
n-Hexacosane

Caution: for Research Use Only

Formula: H₃C(CH₂)₂₄CH₃
MDL Number: MFCD00009354
CAS Number: 630-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR