Dipotassium hexachloropalladate 98%

Supplier: Ambeed
Warning

Synonyms: Potassium hexachloropalladate(IV)

A927526-100MG A927526-1G A927526-250MG A927526-5G
76729-214EA 13 USD
76729-214 76729-216 76729-218 76729-220
Dipotassium hexachloropalladate 98%
Dipotassium hexachloropalladate

Caution: for Research Use Only

Formula: Cl₆K₂Pd
MDL Number: MFCD00043081
CAS Number: 16919-73-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR