N-Acetyl-DL-methionine 98%

Supplier: Ambeed
Warning

Synonyms: Ac-DL-Met-OH

A849460-100G A849460-25G A849460-500G
76728-848EA 19 USD
76728-848 76728-850 76728-852
N-Acetyl-DL-methionine 98%
N-Acetyl-DL-methionine

Caution: for Research Use Only

Formula: C₇H₁₃NO₃S
MDL Number: MFCD00008681
CAS Number: 1115-47-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR