2,3-Naphthalenedialdehyde (NDA) 99%

Supplier: Ambeed
Warning

A770576-100MG A770576-1G A770576-250MG A770576-5G
76728-252EA 42 USD
76728-252 76728-254 76728-256 77602-972
2,3-Naphthalenedialdehyde (NDA) 99%
2,3-Naphthalenedialdehyde (NDA)

Caution: for Research Use Only

Formula: C₁₂H₈O₂
MDL Number: MFCD00016618
CAS Number: 7149-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR