Nitrilotris(methylenephosphonic acid) 50% in water
Supplier: Ambeed
|
Warning
|
Synonyms:
Tris(phosphonomethyl)amine, Nitrilotri(methylphosphonic acid)
A646401-1000G
A646401-100G
A646401-25G
A646401-500G
76727-308EA
17
USD
76727-308
76727-310
76727-312
76727-314
Nitrilotris(methylenephosphonic acid) 50% in water
Nitrilotris(methylenephosphonic acid)
Caution: for Research Use Only
Formula:
C₃H₁₂NO₉P₃ |
MDL Number:
MFCD00002138 CAS Number: 6419-19-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...