Silver trifluoromethanesulfonate 98%

Supplier: Ambeed
Danger

Synonyms: Silver trifluoromethanesulphonate, Silver triflate, Silver trifluoromethanesulfonate, Trifluoromethanesulphonic acid silver salt, Trifluoromethanesulfonic acid silver salt

A634214-10G A634214-500G A634214-1G A634214-25G A634214-5G
103840-300EA 18 USD
103840-300 76727-178 76727-180 77308-020 77308-022 77308-024
Silver trifluoromethanesulfonate 98%
Silver trifluoromethanesulphonate

Caution: for Research Use Only

Formula: CAgF₃O₃S
MW: 256.94 g/mol
Boiling Pt: 162 °C (760 mm Hg)
Melting Pt: 356
Storage Temperature: Ambient
MDL Number: MFCD00013226
CAS Number: 2923-28-6
UN: 1759
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR