Caprylic acid 95%

Supplier: Ambeed
Danger

Synonyms: Octanoic acid, n-Octanoic Acid, Octoic acid

A627710-25G
76727-114EA 6.37 USD
76727-114
Caprylic acid 95%
Caprylic acid

Caution: for Research Use Only

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
MDL Number: MFCD00004429
CAS Number: 124-07-2
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR