1-Benzyl-4-hydroxypiperidine 98%

Supplier: Ambeed
Warning

Synonyms: 1-Benzylpiperidin-4-ol

A612978-100G A612978-10G A612978-25G A612978-500G A612978-5G A612978-1000G
76727-000EA 32 USD
76727-000 76727-002 76727-004 76727-006 76727-008 77669-634
1-Benzyl-4-hydroxypiperidine 98%
1-Benzyl-4-hydroxypiperidine

Caution: for Research Use Only

Formula: C₁₂H₁₇NO
MDL Number: MFCD00006503
CAS Number: 4727-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR