Phenyl bromoacetate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Bromoacetic acid Phenyl ester, Phenyl 2-bromoacetate
A561131-100G
A561131-10G
A561131-5G
A561131-25G
76726-544EA
33
USD
76726-544
76726-546
76726-548
76843-976
Phenyl bromoacetate 97%
Phenyl bromoacetate
Caution: for Research Use Only
Formula:
C₈H₇BrO₂ |
MDL Number:
MFCD00192391 CAS Number: 620-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...