SATA (N-Succinimidyl S-Acetylthioglycolate) 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate
A508892-250MG
A508892-5G
A508892-1G
A508892-100MG
76726-120EA
152
USD
76726-120
76726-122
77568-992
77602-172
SATA (N-Succinimidyl S-Acetylthioglycolate) 98%
SATA (N-Succinimidyl S-Acetylthioglycolate)
Caution: For research use only.
Formula:
C₈H₉NO₅S MW: 231.22 g/mol |
MDL Number:
MFCD00036891 CAS Number: 76931-93-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...