Potassium hexachloroplatinate(IV) 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
Dipotassium hexachloroplatinate
A472623-1G
A472623-250MG
A472623-5G
76725-800EA
54
USD
76725-800
76725-802
77880-028
Potassium hexachloroplatinate(IV) 97%
Potassium hexachloroplatinate(IV)
Caution: for Research Use Only
Formula:
K₂PtCl₆ |
MDL Number:
MFCD00011389 CAS Number: 16921-30-5 UN: 3290 ADR: 6.1,8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...