Potassium carbonate 99%

Supplier: Ambeed
Warning

A442495-100G
76725-484EA 5 USD
76725-484 76725-486
Potassium carbonate 99%
Potassium carbonate

Caution: for Research Use Only

Formula: K₂CO₃
MDL Number: MFCD00011382
CAS Number: 584-08-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR