Ruthenium(III) iodide 98%

Supplier: Ambeed
Danger

A406379-100MG A406379-1G A406379-250MG A406379-5G
76725-128EA 8 USD
76725-128 76725-130 76725-132 76725-134
Ruthenium(III) iodide 98%
Ruthenium(III) iodide

Caution: for Research Use Only

Formula: I₃Ru
MDL Number: MFCD00016316
CAS Number: 13896-65-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR