2-Phenylbenzoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid
A348320-100G
A348320-10G
A348320-500G
A348320-25G
A348320-5G
A348320-1000G
103836-980EA
5
USD
103836-980
76724-414
76724-416
76724-418
76840-058
77306-596
77879-752
2-Phenylbenzoic acid 98%
2-Phenylbenzoic acid
Caution: for Research Use Only
Formula:
C₁₃H₁₀O₂ Melting Pt: 111…113 °C Flash Pt: 343 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002463 CAS Number: 947-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...