N,N'-Bis(2-aminoethyl)-1,3-propanediamine 95%

Supplier: Ambeed
Danger

Synonyms: 3,7-Diaza-1,9-nonanediamine, N1,N1'-(Propane-1,3-diyl)bis(ethane-1,2-diamine), 1,4,8,11-Tetraazaundecane

A339712-10G A339712-1G A339712-25G A339712-5G
76724-312EA 32 USD
76724-312 76724-314 76724-316 76724-318
N,N'-Bis(2-aminoethyl)-1,3-propanediamine 95%
N,N'-Bis(2-aminoethyl)-1,3-propanediamine

Caution: for Research Use Only

Formula: C₇H₂₀N₄
MDL Number: MFCD00008174
CAS Number: 4741-99-5
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR