1,10-Phenanthroline 98%

Supplier: Ambeed
Danger

Synonyms: o-Phenanthroline

A326753-10G A326753-25G A326753-5G A326753-100G
103836-396EA 8 USD
103836-396 76724-064 76839-362 76839-364 77601-470
1,10-Phenanthroline 98%
1,10-Phenanthroline

Caution: for Research Use Only

Formula: C₁₂H₈N₂
Boiling Pt: 365 °C (760 mm Hg)
Melting Pt: 97…102 °C
Flash Pt: 330 °C
Storage Temperature: Ambient
MDL Number: MFCD00011678
CAS Number: 66-71-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR