Tetrasodium diphosphate 99%

Supplier: Ambeed
Warning

Synonyms: Sodium pyrophosphate tetrabasic, Tetrasodium diphosphate, Sodium diphosphate, Sodium pyrophosphate, tetra-Sodium pyrophosphate

A289845-25G
76723-482EA 5 USD
76723-482
Tetrasodium diphosphate 99%
tetra-Sodium diphosphate

Caution: for Research Use Only

Formula: Na₄O₇P₂
MDL Number: MFCD00003513
CAS Number: 7722-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR