8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one 95%

Supplier: Ambeed
Warning

A273593-100G A273593-10G A273593-1G A273593-5G A273593-250MG A273593-25G
76723-094EA 637 USD
76723-094 76723-096 76723-098 76723-100 77668-196 77668-198
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one 95%
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one

Caution: for Research Use Only

Formula: C₁₄H₁₀ClNO
MDL Number: MFCD00800222
CAS Number: 31251-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR