Bis(pinacolato)diboron 98%

Supplier: Ambeed
Warning

Synonyms: 4,4,4',4',5,5,5',5'-Octamethyl-2,2'-bi(1,3,2-dioxaborolane), 4,4,5,5-tetramethyl-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,2-dioxaborolane

A247960-100G A247960-10G A247960-500G A247960-5G A247960-25G
103809-764EA 197 USD
103809-764 76722-558 76722-560 76722-562 76722-564 77090-980
Bis(pinacolato)diboron 98%
Bis(pinacolato)diboron

Caution: for Research Use Only

Formula: C₁₂H₂₄B₂O₄
Boiling Pt: 222.6 °C (760 mm Hg)
Melting Pt: 137…140 °C
Flash Pt: 88.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00799570
CAS Number: 73183-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR