Potassium tripolyphosphate 96%

Supplier: Ambeed
Warning

Synonyms: Potassium triphosphate

A209165-1000G A209165-100G A209165-500G
76721-652EA 42 USD
76721-652 76721-654 76721-656
Potassium tripolyphosphate 96%
Potassium tripolyphosphate

Caution: for Research Use Only

Formula: K₅O₁₀P₃
MDL Number: MFCD00054129
CAS Number: 13845-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR