Potassium tripolyphosphate 96%
Supplier: Ambeed
|
Warning
|
Synonyms:
Potassium triphosphate
A209165-1000G
A209165-100G
A209165-500G
76721-652EA
42
USD
76721-652
76721-654
76721-656
Potassium tripolyphosphate 96%
Potassium tripolyphosphate
Caution: for Research Use Only
Formula:
K₅O₁₀P₃ |
MDL Number:
MFCD00054129 CAS Number: 13845-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...