Methyl cinnamate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-phenylpropenoate, Methyl 3-phenyl-2-propenoate

A202087-1000G A202087-100G A202087-25G A202087-500G
76721-490EA 29 USD
76721-490 76721-492 76721-494 76721-496
Methyl cinnamate 98%
Methyl cinnamate

Caution: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
MDL Number: MFCD00008458
CAS Number: 103-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR