n-Tetracosane 98%

Supplier: Ambeed
Warning

Synonyms: N-tetracosane

A191824-100G A191824-25G A191824-5G
76721-138EA 180 USD
76721-138 76721-140 76721-142
n-Tetracosane 98%
n-Tetracosane

Caution: for Research Use Only

Formula: H₃C(CH₂)₂₂CH₃
MW: 338.66 g/mol
Boiling Pt: 391.3
MDL Number: MFCD00009352
CAS Number: 646-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR