Ruthenium(III) acetylacetonate 98%

Supplier: Ambeed
Danger

Synonyms: Ruthenium (III) 2,4-pentanedionate, Tris(2,4-pentanedionato-O,O')ruthenium (III), Ruthenium (III) tris(2,4-pentanedionato-O,O'), Tris(2,4-pentanedionato)ruthenium(III)

A181230-1G A181230-25G A181230-5G
76720-884EA 21 USD
76720-884 76720-886 76720-888
Ruthenium(III) acetylacetonate 98%
Ruthenium (III) acetylacetonate

Caution: for Research Use Only

Formula: C₁₅H₂₁O₆Ru
MDL Number: MFCD00000030
CAS Number: 14284-93-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR