2',3'-O-Isopropylideneadenosine 98%
Supplier: Ambeed
|
Warning
|
A177625-100G
A177625-10G
A177625-1G
A177625-250MG
A177625-25G
A177625-5G
A177625-500G
103845-584EA
14
USD
103845-584
76720-728
76720-730
76720-732
76720-734
77082-970
77082-972
77879-171
2',3'-O-Isopropylideneadenosine 98%
2',3'-O-Isopropylideneadenosine
Caution: for Research Use Only
Formula:
C₁₃H₁₇N₅O₄ Melting Pt: 221…222 °C Flash Pt: 298.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005756 CAS Number: 362-75-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...